Merck
Droperidol: efficacy and side effects in psychiatric emergencies.
The Journal of clinical psychiatry (1999-11-05)
R A Chambers, B G Druss
ABSTRACT
As admission criteria to inpatient units become more focused on patient safety and behavioral instability, primary treatment often requires use of medications that need to be quick, safe, and effective for control of agitation. This article reviews the evidence that droperidol may serve as the optimal medication for this task. A comprehensive MEDLINE search of English-language literature was conducted using the search term droperidol concerning the use of droperidol in psychiatric emergencies. Cross-referencing of those articles was conducted to include pertinent articles in the non-psychiatric and European literature regarding safety and early development of the drug. As evidenced in the animal and clinical literature, studies demonstrate the efficacy and rapidity of onset of droperidol and its relative safety compared with the most widely used antiagitation drug, haloperidol. Evidence for this use of droperidol is particularly compelling for situations in which intramuscular administration is necessary. Droperidol, while not in widespread use, may prove to be the superior typical neuroleptic for psychiatric emergencies. Increased clinical utilization and study of droperidol for this use is warranted.
Sigma-Aldrich
Droperidol
Droperidol, European Pharmacopoeia (EP) Reference Standard
USP
Droperidol, United States Pharmacopeia (USP) Reference Standard