Merck
  • Mutation of junctophilin type 2 associated with hypertrophic cardiomyopathy.
Mutation of junctophilin type 2 associated with hypertrophic cardiomyopathy.
Journal of human genetics (2007-05-04)
Yoshihisa Matsushita, Toru Furukawa, Hiroshi Kasanuki, Makoto Nishibatake, Yachiyo Kurihara, Atsushi Ikeda, Naoyuki Kamatani, Hiroshi Takeshima, Rumiko Matsuoka
ABSTRACT
Junctophilin subtypes, designated as JPH1 approximately 4, are protein components of junctional complexes and play essential roles in cellular Ca2+ signaling in excitable cells. Knockout mice lacking the cardiac-type Jph2 die of embryonic cardiac arrest, and the mutant cardiac myocytes exhibit impaired formation of peripheral couplings and arrhythmic Ca2+ signaling caused by functional uncoupling between dihydropyridine and ryanodine receptor channels. Based on these observations, we hypothesized that mutations of JPH2 could cause human genetic cardiac diseases. Among 195 Japanese patients (148 index cases and 47 affected family members) with hypertrophic cardiomyopathy (HCM), two heterozygous nonsynonymous nucleotide transitions, G505S and R436C, were newly found in JPH2. When Fisher's exact test was used to compare index cases with HCM to unrelated Japanese healthy controls in the frequencies of mutant alleles, only the G505S mutation showed statistical significance (4/296 HCM patients and 0/472 control individuals, P=0.022). This result was still significant after Bonferroni's correction for multiple comparisons (P=0.044). To the best of our knowledge, this is the first report on JPH2 mutation associated with HCM.
Sigma-Aldrich
Anti-JPH2 antibody produced in rabbit, affinity isolated antibody