Merck
  • Surfactant-modified nanoclay exhibits an antiviral activity with high potency and broad spectrum.
Surfactant-modified nanoclay exhibits an antiviral activity with high potency and broad spectrum.
Journal of virology (2014-01-31)
Jian-Jong Liang, Jiun-Chiou Wei, Yi-Ling Lee, Shan-hui Hsu, Jiang-Jen Lin, Yi-Ling Lin
ABSTRACT
Nanomaterials have the characteristics associated with high surface-to-volume ratios and have been explored for their antiviral activity. Despite some success, cytotoxicity has been an issue in nanomaterial-based antiviral strategies. We previously developed a novel method to fully exfoliate montmorillonite clay to generate the most fundamental units of nanoscale silicate platelet (NSP). We further modified NSP by capping with various surfactants and found that the surfactant-modified NSP (NSQ) was less cytotoxic. In this study, we tested the antiviral potentials of a series of natural-clay-derived nanomaterials. Among the derivatives, NSP modified with anionic sodium dodecyl sulfate (NSQc), but not the pristine clay, unmodified NSP, a silver nanoparticle-NSP hybrid, NSP modified with cationic n-octadecanylamine hydrochloride salt, or NSP modified with nonionic Triton X-100, significantly suppressed the plaque-forming ability of Japanese encephalitis virus (JEV) at noncytotoxic concentrations. NSQc also blocked infection with dengue virus (DEN) and influenza A virus. Regarding the antiviral mechanism, NSQc interfered with viral binding through electrostatic interaction, since its antiviral activity can be neutralized by Polybrene, a cationic polymer. Furthermore, NSQc reduced the lethality of JEV and DEN infection in mouse challenge models. Thus, the surfactant-modified exfoliated nanoclay NSQc may be a novel nanomaterial with broad and potent antiviral activity. Nanomaterials have being investigated as antimicrobial agents, yet their antiviral potential is overshadowed by their cytotoxicity. By using a novel method, we fully exfoliated montmorillonite clay to generate the most fundamental units of nanoscale silicate platelet (NSP). Here, we show that the surfactant-modified NSP (NSQ) is less cytotoxic and that NSQc (NSP modified with sodium dodecyl sulfate) could potently block infection by dengue virus (DEN), Japanese encephalitis virus (JEV), and influenza A virus at noncytotoxic concentrations. For the antiviral mechanism, we find that the electrostatic interaction between the negatively charged NSQc and the positively charged virus particles blocks viral binding. Furthermore, we used mouse challenge models of JEV and DEN to demonstrate the in vivo antiviral potential of NSQc. Thus, NSQc may function as a potent and safe antiviral nanohybrid against several viruses, and our success in synthesizing surfactant-modified NSP with antiviral activity may shed some light on future antiviral development.
Sigma-Aldrich
Triton X-100, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-45
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bentonite
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-102
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
SKU
Pack Size
Availability
Price
Quantity
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard