Merck
  • Low baseline salivary alpha-amylase in drug-naïve patients with short-illness-duration first episode major depressive disorder.
Low baseline salivary alpha-amylase in drug-naïve patients with short-illness-duration first episode major depressive disorder.
Journal of affective disorders (2014-03-04)
Wiesław Jerzy Cubała, Jerzy Landowski
ABSTRACT
Altered monoamine neurotransmission accompanied by hypothalamic-pituitary-adrenal axis dysfunction and autonomic nervous system hyperactivity have been associated with major depressive disorder (MDD). Salivary α-amylase (sAA) is indicative of autonomic activation and reflects central noradrenergic activity. Scarce studies on sAA in MDD produce confounded results and no data is available regarding baseline sAA activity. The basal, non-stimulated sAA activity was studied in this cross-sectional case-control study on 20 non-late-life adult, short-illness-duration first-episode, treatment-naïve MDD patients and in 20 age- and sex-matched healthy controls. Depressed patients showed a basal score in the Hamilton Rating Scale for Depression (HAMD-17) higher than 20. The sAA was significantly lower in depressed individuals as compared to controls (p=0.011). In post hoc analysis significantly lower sAA was present in melancholic MDD (p=0.016) as related to controls whereas no difference was seen between non-melancholic MDD patients and controls. The sAA activity was not significantly correlated neither with duration nor the severity of depressive symptoms as measured by the total HAMD-17 score. The current study is limited by its cross-sectional design, small sample size, and factors related to saliva sampling methodology. Low baseline sAA levels were found in MDD in basal, non-stimulated conditions. The study provides no support for elevated sAA in drug-naïve patients with short-illness-duration first episode MDD. The results support the evidence for decreased central noradrenergic transmission in MDD when sAA activity is considered indicative of central noradrenergic function.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein