Merck
  • Enantioseparation of omeprazole and its metabolite 5-hydroxyomeprazole using open tubular capillary electrochromatography with immobilized avidin as chiral selector.
Enantioseparation of omeprazole and its metabolite 5-hydroxyomeprazole using open tubular capillary electrochromatography with immobilized avidin as chiral selector.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-06-25)
J Olsson, L G Blomberg
ABSTRACT
The present paper demonstrates the enantiomeric separation of omeprazole and its metabolite 5-hydroxyomeprazole performed with open tubular capillary electrochromatography (OT-CEC). The protein avidin was used as the chiral selector. Avidin was immobilized by a Schiffs base type of reaction where the protein was via glutaraldehyde covalently bonded to the amino-modified wall of a fused-silica capillary, 50 microm i.d. Both racemates were baseline resolved. Resolution was 1.9 and 2.3, respectively, using ammonium acetate buffer, pH 5.8, 5% methanol, with UV-detection. These values of resolution using OT-CEC are higher than earlier published results regarding chiral separation of omeprazole and 5-hydroxyomeprazole on packed CEC. The number of theoretical plates also indicated good separation efficiency.
Product Description
Supelco
5-Hydroxyomeprazole, analytical standard