Merck
HPTLC determination of vasicine and vasicinone in Adhatoda vasica.
Phytochemical analysis : PCA (2005-05-11)
Chaitali Das, Rajlakshmi Poi, Ashim Chowdhury
ABSTRACT
A simple high-performance thin-layer chromatographic (HPTLC) method has been developed for the simultaneous determination of the pharmacologically important quinazoline alkaloids vasicine and vasicinone in Adhatoda vasica. The assay combines the separation and quantification of the analytes on silica gel 60 GF254 HPTLC plates with visualisation under UV and scanning at 270 and 281 nm. Using this technique, the alkaloidal content of different parts of the title plant have been determined.