Merck
  • Myogenic effect of SP-1f and SP-1h two novel β3-adrenoceptor (β3-AR) agonists in human colonic circular smooth muscle.
Myogenic effect of SP-1f and SP-1h two novel β3-adrenoceptor (β3-AR) agonists in human colonic circular smooth muscle.
European journal of pharmacology (2013-11-28)
Maria Antonietta Maselli, Piero Trisolini, Antonia Ignazzi, Maria Notarnicola, Dionigi Lorusso, Francesco Pezzolla, Dominga Iacobazzi, Antonio Scilimati, Maria Grazia Perrone
ABSTRACT
The effect of two novel β3-adrenoceptor (β3-AR) agonists SP-1f and SP-1h on human colon circular smooth muscle contractility and β3-AR mRNA expression have been determined. β3-AR is ascertained co-participates to the control of the gut motility. Isometric tension on human colon muscle strips was measured in response to increasing concentrations of SP-1f, SP-1h and (-)-isoprenaline, alone and in the presence of Betaxolol, ICI 11,855 and SR 59230A (β1-, β2- and β3-AR antagonists, respectively). (-)-Isoprenaline concentration-dependently relaxed circular muscle strips with an EC50=0.32±0.06μM. Such an effect was antagonized either by the contemporaneously presence of Betaxolol and ICI 11,855 [(-)-isoprenaline EC50=1.75±0.35μM, pKB=7.88±0.10] or by Betaxolol, ICI 11,855 and SR 59230A [(-)-isoprenaline EC50=3.49±0.38μM, pKB=8.51±0.14]. Besides, SP-1f and SP-1h concentration-dependently relaxed circular muscle strips with an EC50=0.35±0.07μM and 0.45±0.12μM, respectively. These values remained unchanged by blocking the β1- and β2-AR. The presence of SR 59230A antagonized the relaxing effect of SP-1f (EC50=3.51±0.94μM, pKB=8.93±0.16) and did not modify the SP-1h relaxing potency. In colon circular smooth muscle and in mucosa, β3-AR mRNA expression levels were found to be 0.39±0.70 and 0.26±0.12 (P<0.05), respectively. Such results provide further evidence of the β3-adrenoceptor functional role in the human colon and the crucial contribution of SP-1f to the control of the gut dysmotility.
Sigma-Aldrich
SR 59230A, ≥98% (HPLC), powder