Merck
  • [New derivatives of azobenzene for the directed modification of proteins].
[New derivatives of azobenzene for the directed modification of proteins].
Bioorganicheskaia khimiia (2009-11-17)
Le Tkhi Khien, B Shirling, A Iu Riazanova, T S Zatsepin, E M Volkov, E A Kubareva, T I Velichko, A Pingoud, T S Oretskaia
ABSTRACT
Derivatives of azobenzene which contained a maleimide group in one of the benzene rings (for binding to a protein cysteine residue) and maleimide, hydroxyl, or carboxyl substitutes in another benzene ring were synthesized. The reactivity of these compounds towards a cysteine residue of a protein and their optical properties in a free state and after their attachment to the mutant forms of the SsoII restriction endonuclease were studied.
Sigma-Aldrich
4-Aminotoluene-3-sulfonic acid, 99%