Merck
  • Flexible protocol for the chemo- and regioselective building of pyrroles and pyrazoles by reactions of Danishefsky's dienes with 1,2-diaza-1,3-butadienes.
Flexible protocol for the chemo- and regioselective building of pyrroles and pyrazoles by reactions of Danishefsky's dienes with 1,2-diaza-1,3-butadienes.
Organic letters (2008-04-23)
Orazio A Attanasi, Gianfranco Favi, Paolino Filippone, Gianluca Giorgi, Fabio Mantellini, Giada Moscatelli, Domenico Spinelli
ABSTRACT
The versatility of the Mukaiyama-Michael-type addition/heterocyclization of Danishefsky's diene with 1,2-diaza-1,3-butadienes was applied to the synthesis of both 4 H-1-aminopyrroles and 4,5 H-pyrazoles. Thus, the same reagents furnished different types of highly functionalized azaheterocycles essentially depending on their structure: as a matter of fact, R1 = COOR or CONR 2 differently affects the acidity of the proton at the adjacent carbon. An unexpected formation of 5 H-1-aminopyrroles from the reactions carried out in water was also observed.
Sigma-Aldrich
trans-1-Methoxy-3-trimethylsiloxy-1,3-butadiene, 95%