Merck
  • Proteomic analysis reveals that Adh1p is involved in a synergistic fluconazole and tetrandrine mechanism against Candida albicans.
Proteomic analysis reveals that Adh1p is involved in a synergistic fluconazole and tetrandrine mechanism against Candida albicans.
Die Pharmazie (2014-01-10)
Xiao-Li Zhang, Ge-Hua Zhang, Zhi-Yuan Wang, Hui Guo, Lai-Qiang Gao, Si Wu, Yan-Jun Song, Hong Zhang
ABSTRACT
We previously showed that Adh1p participates in fluconazole (FLC) resistance in Candida albicans through a mechanism that may involve efflux pumps. We also found that the concomitant use of tetrandrine (TET) and FLC provided a synergistic action against C. albicans and that the mechanism of action could be related to inhibition of a drug efflux system. To determine whether Adh1p participates in the synergistic antifungal activity of TET against C. albicans, we performed a comparative proteomic study comparing cells treated with FLC and/or TET in FLC-sensitive CA-3 and untreated control cells. Proteins were analyzed using two-dimensional polyacrylamide gel electrophoresis (2-D PAGE), and differentially expressed proteins were identified through matrix-assisted laser desorption/ionization time-of-flight (MALDI TOF/TOF) mass spectrometry. The resulting data were searched against a C. albicans protein database. Our analyses identified six differentially expressed proteins; four (Eno1p, Adh1p, Slb1p, and Tdh1p) were down-regulated, and two (Xyl2p, and Cdc19p) were up-regulated. The Adh1p mRNA levels were consistent with the Adh1p protein levels in all of the groups. The results suggest that Adh1p participates in the synergistic antifungal activity of TET against C. albicans.
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Supelco
Tetrandrine, analytical standard, for drug analysis
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard