Merck
  • A retrospective assessment of the incidence of respiratory depression after neuraxial morphine administration for postcesarean delivery analgesia.
A retrospective assessment of the incidence of respiratory depression after neuraxial morphine administration for postcesarean delivery analgesia.
Anesthesia and analgesia (2013-11-22)
Theresa R Crowgey, Jennifer E Dominguez, Cathleen Peterson-Layne, Terrence K Allen, Holly A Muir, Ashraf S Habib
ABSTRACT
Respiratory depression can occur after neuraxial morphine administration. In the obstetric population, there are little data on respiratory depression after neuraxial morphine administration in women undergoing cesarean delivery. In this single-center, retrospective study in 5036 obstetric patients (mean body mass index = 34 kg/m) who underwent cesarean delivery and received neuraxial morphine, we did not identify any instances of respiratory depression requiring naloxone administration or rapid response team involvement. Therefore, the upper 95% confidence limit for respiratory depression in our study is 0.07% (1 event per 1429 cases).
Product Description
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®