Merck
  • Sesamin ameliorates doxorubicin-induced cardiotoxicity: involvement of Sirt1 and Mn-SOD pathway.
Sesamin ameliorates doxorubicin-induced cardiotoxicity: involvement of Sirt1 and Mn-SOD pathway.
Toxicology letters (2013-11-12)
Suwen Su, Qian Li, Yi Liu, Chen Xiong, Junxia Li, Rong Zhang, Yujie Niu, Lijuan Zhao, Yongli Wang, Huicai Guo
ABSTRACT
Oxidative stress caused by doxorubicin (DOX) is believed to be a major underlying molecular mechanism of DOX-induced cardiotoxicity. Sesamin (Ses), an active component extracted from sesame seeds, exhibits antioxidative and anti-inflammatory effects. In the present study, possible protective mechanisms of Ses on DOX-induced cardiotoxicity were investigated in rats and cultured H9C2 cells. We demonstrated that Ses exhibits a significant protective effect on cardiac tissue in animal and cell models of DOX-induced cardiac injury. Moreover, Ses can ameliorate DOX-induced oxidative stress and mitochondrial damage. Further studies suggested that Ses is able to up-regulate the protein expression of Mn-SOD in normal rats and to restore the decreased expression of Mn-SOD in DOX-induced cardiac injury rats. Exposure to Ses or DOX alone slightly increased the protein expression of Sirt1; however, a more remarkable increase in Sirt1 protein level was detected in the Ses+DOX group. Treatment with a pan-sirtuin inhibitor (nicotinamide) or a Sirt1-specific inhibitor (EX-527) partially antagonised the effect of Ses on DOX-induced mitochondrial damage and completely abolished the effect of Ses on Mn-SOD expression. These findings indicate that the protective mechanisms of Ses on DOX-induced cardiotoxicity are involved in the alleviation of oxidative stress injury and Mn-SOD dysfunction, partially via the activation of Sirt1.
Product Description
Supelco
Sesamin, analytical standard
Pricing and availability is not currently available.