Merck
Allantoin as a solid phase adsorbent for removing endotoxins.
Journal of chromatography. A (2013-09-05)
Vincent Vagenende, Tim-Jang Ching, Rui-Jing Chua, Pete Gagnon
ABSTRACT
In this study we present a simple and robust method for removing endotoxins from protein solutions by using crystals of the small-molecule compound 2,5-dioxo-4-imidazolidinyl urea (allantoin) as a solid phase adsorbent. Allantoin crystalline powder is added to a protein solution at supersaturated concentrations, endotoxins bind and undissolved allantoin crystals with bound endotoxins are removed by filtration or centrifugation. This method removes an average of 99.98% endotoxin for 20 test proteins. The average protein recovery is ∼80%. Endotoxin binding is largely independent of pH, conductivity, reducing agent and various organic solvents. This is consistent with a hydrogen-bond based binding mechanism. Allantoin does not affect protein activity and stability, and the use of allantoin as a solid phase adsorbent provides better endotoxin removal than anion exchange, polymixin affinity and biological affinity methods for endotoxin clearance.
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Supelco
Allantoin, analytical standard
Allantoin, European Pharmacopoeia (EP) Reference Standard