Merck
  • Paralog-selective Hsp90 inhibitors define tumor-specific regulation of HER2.
Paralog-selective Hsp90 inhibitors define tumor-specific regulation of HER2.
Nature chemical biology (2013-09-03)
Pallav D Patel, Pengrong Yan, Paul M Seidler, Hardik J Patel, Weilin Sun, Chenghua Yang, Nanette S Que, Tony Taldone, Paola Finotti, Ralph A Stephani, Daniel T Gewirth, Gabriela Chiosis
ABSTRACT
Although the Hsp90 chaperone family, comprised in humans of four paralogs, Hsp90α, Hsp90β, Grp94 and Trap-1, has important roles in malignancy, the contribution of each paralog to the cancer phenotype is poorly understood. This is in large part because reagents to study paralog-specific functions in cancer cells have been unavailable. Here we combine compound library screening with structural and computational analyses to identify purine-based chemical tools that are specific for Hsp90 paralogs. We show that Grp94 selectivity is due to the insertion of these compounds into a new allosteric pocket. We use these tools to demonstrate that cancer cells use individual Hsp90 paralogs to regulate a client protein in a tumor-specific manner and in response to proteome alterations. Finally, we provide new mechanistic evidence explaining why selective Grp94 inhibition is particularly efficacious in certain breast cancers.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Purine, 98%