Merck
  • The effect of melatonin on plasma markers of inflammation and on expression of nuclear factor-kappa beta in acetic acid-induced colitis in the rat.
The effect of melatonin on plasma markers of inflammation and on expression of nuclear factor-kappa beta in acetic acid-induced colitis in the rat.
Digestive diseases and sciences (2013-08-09)
Hayam G Sayyed, Rumi J Jaumdally, Naglaa K Idriss, Dalia A El Sers, Andrew Blann
ABSTRACT
Melatonin may be involved in gastrointestinal tract physiology and could affect inflammation-related gastrointestinal disorders. Rat models of ulcerative colitis imply melatonin is beneficial. To determine potential pathophysiological mechanisms, we assessed colonic nuclear factor-kappa beta expression and measured serum levels of pentraxin-3, lipid peroxides, and total thiols in an acetic acid model of this disease. Thirty rats were divided into five groups: a control group, an acetic acid-induced colitis group, a group treated with melatonin before colitis induction, a group treated short-term after colitis induction, and a group treated long-term after colitis induction. After four weeks, blood samples were taken for measurement of pentraxin-3, lipid peroxide, and total thiols. Sections of the colon were taken for histopathological examination and immunohistochemical detection of nuclear factor-kappa beta expression. Melatonin administration reduced nuclear factor-kappa beta immunohistochemical expression, reduced serum levels of lipid peroxide and pentraxin-3, and maintained serum levels of total thiols. However, in long-term treatment the protective effect of melatonin was not as marked. Melatonin is effective in prevention and short-term treatment of the inflammatory process in acetic-acid induced colitis whereas the benefit of long-term treatment is unclear. Benefit may be linked to protection mechanisms against inflammatory processes by inhibiting the nuclear factor-kappa beta and conserving endogenous antioxidant reserves of total thiols, thus reducing the level of colonic damage possibly caused by lipid peroxides.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology