Merck
  • Evaluation of the enantioselectivity of glycogen-based synergistic system with amino acid chiral ionic liquids as additives in capillary electrophoresis.
Evaluation of the enantioselectivity of glycogen-based synergistic system with amino acid chiral ionic liquids as additives in capillary electrophoresis.
Journal of chromatography. A (2013-08-06)
Qi Zhang, Yingxiang Du
ABSTRACT
In this paper, two novel amino acid chiral ILs, tetramethylammonium-l-arginine (TMA-l-Arg) and tetramethylammonium-l-aspartic acid (TMA-l-Asp), were applied for the first time in CE enantioseparation to evaluate their potential synergistic effect with glycogen as chiral selector. As observed, significantly improved separation of tested enantiomers were obtained in the chiral ILs/glycogen synergistic systems compared to the single glycogen separation system. Several primary parameters affecting the enantioseparation, such as amino acid ILs (AAILs) concentration, glycogen concentration and buffer pH, were systematically investigated. An achiral tetramethylammonium hydroxide ionic liquid (TMA-OH) modified separation system was also evaluated to validate the superiority of the novel chiral ILs/glycogen synergistic systems. To further optimize the overall synergistic systems, the effect of three other parameters, including buffer concentration, applied voltage and capillary temperature were simultaneously analyzed by a central composite design (CCD), and excellent enantioseparations were achieved with the optimized parameters. The results indicate that the application of chiral ILs/glycogen synergistic systems is a promising way in chiral separation science.
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
Glycogen from oyster
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)