Merck
  • 3D pure inorganic framework based on polymolybdovanadate possessing photoelectric properties.
3D pure inorganic framework based on polymolybdovanadate possessing photoelectric properties.
Dalton transactions (Cambridge, England : 2003) (2013-07-19)
Fengyan Li, Fanzhi Meng, Lifang Ma, Lin Xu, Zhixia Sun, Qiao Gao
ABSTRACT
The first 3D pure inorganic framework based on heteropolymolybdate anion [V2Mo16O58](10-) has been synthesized and its photoelectric properties demonstrated.
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis