Merck
  • Observation of the dynamic germination of single bacterial spores using rapid Raman imaging.
Observation of the dynamic germination of single bacterial spores using rapid Raman imaging.
Journal of biomedical optics (2013-07-12)
Lingbo Kong, Peter Setlow, Yong-qing Li
ABSTRACT
The dynamics of bacterial spore germination were successfully observed using a fast Raman imaging system, in combination with real-time phase contrast microscopy. By using a multifocus scan scheme, the spontaneous Raman-scattering imaging acquisition speed was increased to ~30 s per frame while maintaining diffraction-limited resolution, which allowed monitoring of the dynamics of spore germination on a time scale of tens of seconds to a few minutes. This allowed simultaneous gathering of rich spatial distribution information on different cellular components including time-lapse molecular images of Ca-dipicolinic acid, protein, and nucleic acid during germination of single bacterial spores for the periods of 30 to 60 min.
Product Description
Supelco
2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%