Merck
  • Performance of passive samplers for monitoring estuarine water column concentrations: 1. Contaminants of concern.
Performance of passive samplers for monitoring estuarine water column concentrations: 1. Contaminants of concern.
Environmental toxicology and chemistry (2013-07-09)
Monique M Perron, Robert M Burgess, Eric M Suuberg, Mark G Cantwell, Kelly G Pennell
ABSTRACT
Contaminants enter marine and estuarine environments and pose a risk to human and ecological health. Recently, passive sampling devices have been utilized to estimate dissolved concentrations of contaminants of concern (COCs), such as polycyclic aromatic hydrocarbons (PAHs) and polychlorinated biphenyls (PCBs). In the present study, the performance of 3 common passive samplers was evaluated for sampling PAHs and PCBs at several stations in the temperate estuary Narragansett Bay, Rhode Island, USA. Sampler polymers included polyethylene (PE), polydimethylsiloxane (PDMS)-coated solid-phase microextraction (SPME) fibers, and polyoxymethylene (POM). Dissolved concentrations of each contaminant were calculated using measured sampler concentrations adjusted for equilibrium conditions with performance reference compounds (PRCs) and chemical-specific partition coefficients derived in the laboratory. Despite differences in PE and POM sampler concentrations, calculated total dissolved concentrations ranged from 14 ng/L to 93 ng/L and from 13 pg/L to 465 pg/L for PAHs and PCBs, respectively. Dissolved concentrations of PAHs were approximately 3 times greater based on POM compared to PE, while dissolved concentrations of PCBs based on PE were approximately 3 times greater than those based on POM. Concentrations in SPME were not reported due to the lack of detectable chemical in the amount of PDMS polymer deployed. Continued research is needed to improve and support PE and POM use for the routine monitoring of COCs. For example, a better understanding of the use of PRCs with POM is critically needed.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)