Merck
  • Poly(N-isopropylacrylamide) hydrogels with interpenetrating multiwalled carbon nanotubes for cell sheet engineering.
Poly(N-isopropylacrylamide) hydrogels with interpenetrating multiwalled carbon nanotubes for cell sheet engineering.
Biomaterials (2013-07-06)
Yu-Shuan Chen, Pei-Chun Tsou, Jem-Mau Lo, Hsieh-Chih Tsai, Yan-Zhen Wang, Ging-Ho Hsiue
ABSTRACT
Hydrogels have been developed as artificial extracellular matrixes (ECMs) to mimic native tissue microenvironments for various applications. Unfortunately, poly(N-isopropylacrylamide) (PNIPAAM)-based hydrogels are not suitable for cell culturing and cell sheet preparation. Carbon nanotubes (CNTs), with their mechanical strength and electrical conductivity, have been considered as additives to increase the applicability of hydrogels to cell encapsulation and advance cardiac electrophysiological functions. A simple method for fabrication of PNIPAAM hydrogels interpenetrated with multiwalled CNTs (MWCNTs) as substrates for cell sheet preparation is reported. The results demonstrate that PNIPAAM hydrogels with interpenetrating MWCNTs still exhibit thermosensitive behavior. It is also found that epithelial Madin-Darby canine kidney (MDCK) cells can only attach and proliferate on MWCNT-interpenetrated PNIPAAM hydrogels. Furthermore, the PNIPAAM hydrogels with MWCNTs possess higher elastic moduli and hydrophobicities than those without MWCNTs, suggesting these two characteristics are necessary for the cells to attach to the hydrogel surfaces. Moreover, cell sheets can only be harvested from PNIPAAM hydrogels with MWCNTs because of their high ratio of cell attachment. Thus, this simple method provides sufficient mechanical strength to PNIPAAM hydrogels so that anchorage-dependent cells can be cultivated and provides a superior system for preparing cell sheets.
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-7491957-100MG100 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,042.40
EN300-7491957-250MG250 mg
Ships in 14 days - from Aldrich Partner.
NOK 2,700.20
EN300-7491957-500MG500 mg
Ships in 14 days - from Aldrich Partner.
NOK 3,956.00
EN300-7491957-1G1 g
Ships in 14 days - from Aldrich Partner.
NOK 4,927.75
EN300-7491957-50MG50 mg
Ships in 14 days - from Aldrich Partner.
NOK 7,484.20
EN300-7491957-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
NOK 9,786.50
EN300-7491957-5G5 g
Ships in 14 days - from Aldrich Partner.
NOK 17,889.40
EN300-7491957-10G10 g
Ships in 14 days - from Aldrich Partner.
NOK 34,095.20