- Fast mass spectrometry detection of tryptophan-containing peptides and proteins by reduction with pyridine-borane.
Fast mass spectrometry detection of tryptophan-containing peptides and proteins by reduction with pyridine-borane.
Analytical biochemistry (2013-05-28)
T Miron, M Wilchek
PMID23707716
ABSTRACT
In this note, we describe a method devised to detect, by means of mass spectrometry (MS), tryptophan-containing peptides and proteins using pyridine-borane. This reagent selectively reduces tryptophan residues, converting them to 2,3-dihydro-tryptophan, thereby enabling quantitation of tryptophans.
MATERIALS