Merck
  • Adalimumab combined with ciprofloxacin is superior to adalimumab monotherapy in perianal fistula closure in Crohn's disease: a randomised, double-blind, placebo controlled trial (ADAFI).
Adalimumab combined with ciprofloxacin is superior to adalimumab monotherapy in perianal fistula closure in Crohn's disease: a randomised, double-blind, placebo controlled trial (ADAFI).
Gut (2013-03-26)
Pieter Dewint, Bettina E Hansen, Elke Verhey, Bas Oldenburg, Daniel W Hommes, Marieke Pierik, Cyriel I J Ponsioen, Hendrik M van Dullemen, Maurice Russel, Ad A van Bodegraven, C Janneke van der Woude
ABSTRACT
To assess whether a combination of adalimumab and ciprofloxacin is superior to adalimumab alone in the treatment of perianal fistulising Crohn's disease (CD). Randomised, double-blind, placebo controlled trial in eight Dutch hospitals. In total, 76 CD patients with active perianal fistulising disease were enrolled. After adalimumab induction therapy (160/80 mg week 0, 2), patients received 40 mg every other week together with ciprofloxacin 500 mg or placebo twice daily for 12 weeks. After 12 weeks, adalimumab was continued. Follow-up was 24 weeks. Primary endpoint (clinical response) was defined as 50% reduction of fistulas from baseline to week 12. Secondary endpoints included remission (closure of all fistulas), Perianal Crohn's Disease Activity Index, Crohn's Disease Activity Index (CDAI) and Inflammatory Bowel Disease Questionnaire (IBDQ). Clinical response was observed in 71% of patients treated with adalimumab plus ciprofloxacin and in 47% treated with adalimumab plus placebo (p=0.047). Likewise, remission rate at week 12 was significantly higher (p=0.009) in the combination group (65%) compared with adalimumab plus placebo (33%). Combination treatment was associated with a higher mean CDAI change and mean IBDQ change at week 12 (p=0.005 and p=0.009, respectively). At week 24, no difference in clinical response between the two treatment groups was observed (p=0.22). No difference in safety issues was observed. Combination therapy of adalimumab and ciprofloxacin is more effective than adalimumab monotherapy to achieve fistula closure in CD. However, after discontinuation of antibiotic therapy, the beneficial effect of initial coadministration is not maintained. ClinicalTrials.gov Identifier: NCT00736983.
Product Description
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material