Merck
Asymmetric synthesis of both enantiomers of arteludovicinolide A.
Organic letters (2013-06-26)
Andreas Kreuzer, Sabine Kerres, Thomas Ertl, Hannelore Rücker, Sabine Amslinger, Oliver Reiser
ABSTRACT
The first total synthesis of either enantiomer of Arteludovicinolide A and their biological evaluation is reported, featuring a new strategy for the asymmetric construction of γ-butyrolactones with stereogenic side chains in the 4-position. Starting from the renewable resource methyl 2-furoate, the sesquiterpene lactone was synthesized in 9 steps and 4.8% overall yield via an asymmetric cyclopropanation and two diastereoselective nucleophile additions making use of a donor-acceptor-cyclopropane-lactonization cascade. At noncytotoxic concentrations (≤10 μM) (+)-1 was found to have a 15 times higher anti-inflammatory activity (4.87 ± 1.1 μM) than previously reported for concentrations of ≥45 μM.
Sigma-Aldrich
Methyl 2-furoate, 98%