Merck
  • Palladium(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
Palladium(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
Organic letters (2011-07-29)
Björn Nelson, Wolf Hiller, Annett Pollex, Martin Hiersemann
ABSTRACT
Scope and limitations of the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes have been studied. In situ NMR experiments indicate a challenging competition between various reaction pathways. A careful balance between substrate structure, nature of the precatalyst, and reaction conditions was required to gain access to a useful building block for sesquiterpene total synthesis.
Sigma-Aldrich
1,5-Hexadiene, 97%