Merck
  • Direct introduction of large-volume urine samples into an on-line immunoaffinity sample pretreatment-capillary gas chromatography system.
Direct introduction of large-volume urine samples into an on-line immunoaffinity sample pretreatment-capillary gas chromatography system.
Analytical chemistry (1991-11-01)
A Farjam, J J Vreuls, W J Cuppen, U A Brinkman, G J de Jong
ABSTRACT
An immunoaffinity precolumn containing immobilized antibodies raised against the synthetic steroid hormone beta-19-nortestosterone, has been used for the automated sample pretreatment of urine samples containing beta-19-nortestosterone or the related steroids norethindrone and norgestrel. The sample pretreatment system was coupled on-line to a capillary GC. The on-line connection between the immunoaffinity precolumn and the capillary GC was realized with an interface that consisted of a 10 mm X 2 mm i.d. reversed-phase pre-column and a diphenyltetramethyldisilazane-deactivated GC retention gap. After preconcentration on the immunoaffinity precolumn the analytes were eluted and reconcentrated on the reversed-phase precolumn. Subsequently, this precolumn was desorbed with 75 microL of ethyl acetate, which was directly introduced into the retention gap by using partially concurrent solvent evaporation. The system allows the automated pretreatment and GC analysis of 5-25-mL urine samples for the ppt-level determination of 19-norsteroids. The general applicability and potential of on-line immunoaffinity-capillary GC systems are discussed.
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)