Merck
  • [125I-Tyr1]biphalin binding to opioid receptors of rat brain and NG108-15 cell membranes.
[125I-Tyr1]biphalin binding to opioid receptors of rat brain and NG108-15 cell membranes.
Life sciences (1998-05-07)
J Slaninova, S M Appleyard, A Misicka, A W Lipkowski, R J Knapp, S J Weber, T P Davis, H I Yamamura, V J Hruby
ABSTRACT
Mono iodinated analogues of biphalin [(Tyr-D-Ala-Gly-Phe-NH-)2], both nonradioactive [I-Tyr1]biphalin and radioactive [125I-Tyr1]biphalin have been synthesized. The radioligand binding profiles of these compounds for two types of tissues, rat brain membranes, and NG108-15 cell membranes were identical to the parent biphalin. This is additional evidence for the hypothesis that biphalin behaves like a monomeric ligand and that only one intact tyrosine is necessary for high biological activity. The second tyrosine could be used for successful radioiodination which may greatly simplify biochemical and pharmacological studies of biphalin. The results of receptor binding studies show that the binding of both biphalin and [I-Tyr1]biphalin to the delta and mu opioid receptors are not independent. [125I-Tyr1]Biphalin binds to delta receptors as shown in NG108-15 cell membranes. Nevertheless, [125I]biphalin binding to delta receptors in rat brain membranes was hardly evident and mu receptor binding predominated or at least was much more readily detectable in this preparation.