Merck
  • Susceptibility testing of Dientamoeba fragilis ATCC 30948 with iodoquinol, paromomycin, tetracycline, and metronidazole.
Susceptibility testing of Dientamoeba fragilis ATCC 30948 with iodoquinol, paromomycin, tetracycline, and metronidazole.
Antimicrobial agents and chemotherapy (1994-05-01)
F T Chan, M X Guan, A M Mackenzie, F Diaz-Mitoma
ABSTRACT
Susceptibility testing was performed on Dientamoeba fragilis ATCC 30948 in a dixenic culture with Klebsiella pneumoniae and Bacteroides vulgatus. D. fragilis was cocultured with the bacteria in TYGM-9 medium (ATCC medium 1171). The activities of antiparasitic drugs were assessed by counting viable D. fragilis trophozoites with a hemacytometer by trypan blue exclusion. The minimal amebicidal concentrations of the following four drugs were determined: iodoquinol at 128 micrograms/ml, paromomycin at 16 micrograms/ml, tetracycline (questionably) at 32 micrograms/ml, and metronidazole at 32 micrograms/ml.
Sigma-Aldrich
5,7-Diiodo-8-hydroxyquinoline, 95%