Merck
  • Protective activity of hamamelitannin on cell damage of murine skin fibroblasts induced by UVB irradiation.
Protective activity of hamamelitannin on cell damage of murine skin fibroblasts induced by UVB irradiation.
Journal of dermatological science (1995-07-01)
H Masaki, T Atsumi, H Sakurai
ABSTRACT
The protective activities of hamamelitannin (2',5-di-O-galloyl-hamamelose) in Hamamelis virginiana L. and its related compound, gallic acid, on damaged murine skin fibroblasts induced by UVB irradiation were investigated. In order to exclude the UV absorbing effect of the compounds, the protection study was performed such that the fibroblasts were pretreated with hamamelitannin or gallic acid for 24 h before UVB irradiation. At 200 microM concentration, hamamelitannin gave the higher survival of 72.6 +/- 0.4% in comparison with that of gallic acid (35.5 +/- 1.0%), while UVB absorbers such as 2-ethylhexyl p-methoxycinnamate and hexylbenzoate did not show such protection. The scavenging activities of hamamelitannin and gallic acid against active oxygens such as superoxide anion radicals, hydroxyl radicals and singlet oxygens were evaluated using electron spin resonance (ESR-spin trapping method). Hamamelitannin and gallic acid showed potent scavenging activities against all active oxygens tested. Furthermore, the association of hamamelitannin to fibroblasts was examined by comparing it with that of gallic acid, and the following results were obtained: (1) hamamelitannin reduces the reaction rate of liposome entrapped-nitroblue tetrazolium (NBT) with external superoxide anions, and (2) several glycosides associate with fibroblasts. From these results, it was concluded that hamamelitannin protects murine fibroblasts against external active oxygens by associating with the cell surface through its sugar moiety.
Sigma-Aldrich
Hamamelitannin, ≥98.0% (HPLC)