Merck
Non-specific esterases in partly mineralized bovine enamel.
Acta odontologica Scandinavica (1990-10-01)
D Moe, S Kirkeby
ABSTRACT
Activity for non-specific esterase was demonstrated in the matrix of developing bovine enamel with alpha-naphthyl acetate and 5-bromoindoxyl acetate as the esterase substrates. By use of high-performance liquid chromatography gel filtration, ion-exchange chromatography, and electrophoresis three esterases were shown to be present in the enamel matrix. The enzymes showed highest activity at pH 6.5-7.5. In sections a strong reaction was observed in the secretory ameloblasts. The esterases may be proteolytic enzymes that participate in the degradation of the matrix proteins.
Sigma-Aldrich
5-Bromoindoxyl acetate, 98%