Merck
  • O6-methylguanine induces altered proteins at the level of transcription in human cells.
O6-methylguanine induces altered proteins at the level of transcription in human cells.
Nucleic acids research (2010-08-13)
John A Burns, Kristian Dreij, Laura Cartularo, David A Scicchitano
ABSTRACT
O(6)-Methylguanine (O(6)-meG), which is produced in DNA following exposure to methylating agents, instructs human RNA polymerase II to mis-insert bases opposite the lesion during transcription. In this study, we examined the effect of O(6)-meG on transcription in human cells and investigated the subsequent effects on protein function following translation of the resulting mRNA. In HEK293 cells, O(6)-meG induced incorporation of uridine or cytidine in nascent RNA opposite the adduct. In cells containing active O(6)-alkylguanine-DNA alkyltransferase (AGT), which repairs O(6)-meG, 3% misincorporation of uridine was observed opposite the lesion. In cells where AGT function was compromised by addition of the AGT inhibitor O(6)-benzylguanine, ∼ 58% of the transcripts contained a uridine misincorporation opposite the lesion. Furthermore, the altered mRNA induced changes to protein function as demonstrated through recovery of functional red fluorescent protein (RFP) from DNA coding for a non-fluorescent variant of RFP. These data show that O(6)-meG is highly mutagenic at the level of transcription in human cells, leading to an altered protein load, especially when AGT is inhibited.
Sigma-Aldrich
6-O-Methylguanine, 97%
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR