Merck
  • Diastereoselective formation of a dicopper(i) helicate with a chiral tetradentate pyridylthiazole ligand.
Diastereoselective formation of a dicopper(i) helicate with a chiral tetradentate pyridylthiazole ligand.
Chemical communications (Cambridge, England) (2009-04-01)
Chui-Shan Tsang, Ho-Lun Yeung, Wing-Tak Wong, Hoi-Lun Kwong
ABSTRACT
Reaction of a pinene-based pyridylthioamide with 1,4-dibromo-2,3-butanedione in refluxing methanol yielded a new chiral pyridylthiazole ligand L which forms a dinuclear double-stranded helicate with Cu(i) ions; this helicate has opposite helical chirality when compared with its quaterpyridine analogue.
Sigma-Aldrich
1,4-Dibromo-2,3-butanedione, 99%