Merck
  • Microbial biodegradation and toxicity of vinclozolin and its toxic metabolite 3,5-dichloroaniline.
Microbial biodegradation and toxicity of vinclozolin and its toxic metabolite 3,5-dichloroaniline.
Journal of microbiology and biotechnology (2008-03-01)
Jung-Bok Lee, Ho-Yong Sohn, Kee-Sun Shin, Jong-Sik Kim, Min-Sub Jo, Chun-Pyo Jeon, Jong-Ok Jang, Jang-Eok Kim, Gi-Seok Kwon
ABSTRACT
Vinclozolin, an endocrine disrupting chemical, is a chlorinated fungicide widely used to control fungal diseases. However, its metabolite 3,5-dichloroaniline is more toxic and persistent than the parent vinclozolin. For the biodegradation of vinclozolin, vinclozolin- and/or 3,5-dichloroaniline-degrading bacteria were isolated from pesticide-polluted agriculture soil. Among the isolated bacteria, a Rhodococcus sp. was identified from a 16S rDNA sequence analysis and named Rhodococcus sp. T1-1. The degradation ratios for vinclozolin or 3,5- dichloroaniline in a minimal medium containing vinclozolin (200 microg/ml) or 3,5-dichloroaniline (120 microg/ml) were 90% and 84.1%, respectively. Moreover, Rhodococcus sp. T1-1 also showed an effective capability to biodegrade dichloroaniline isomers on enrichment cultures in which they were contained. Therefore, these results suggest that Rhodococcus sp. T1-1 can bioremediate vinclozolin as well as 3,5-dichloroaniline.
Sigma-Aldrich
3,5-Dichloroaniline, ≥97%