- Urinary excretion of dietary contaminants in horses.
Urinary excretion of dietary contaminants in horses.
Presence of drugs is completely prohibited in post racing urine samples by most of racing and competition authorities, even if environmental contamination might occur. To assess the daily dose of several contaminants absorbed through the diet that would result in detectable concentrations in urine. Caffeine, theobromine, theophylline, atropine, scopolamine, bufotenine, DMT or morphine were administered orally to 6 horses, in different dosages, for 3 days before their urine was sampled for regular anti-doping tests. Theobromine, theophylline, bufotenine and morphine daily intake >10 mg, 2 mg, 10 mg and 200 microg, respectively, by a performance horse, were found to result in detectable urinary concentrations. At the 2 tested doses, atropine (5 and 15 mg) and dimethyltryptamine (3 and 10 mg) were not detected in urine. For caffeine and scopolamine, even the lowest dosage tested (5 mg/horse/day and 2 mg/horse/day respectively) induced detectable concentrations of the molecule in urine. Horses fed dietary contaminants, even at level much below the effective dosage, may be positive to antidoping urine analysis. Further research is needed to gain more confident results on a daily safe intake for caffeine and scopolamine. Selection of feed materials appears to be of great importance to prevent non voluntary positive result to anti-doping tests.