Merck
  • Effect of clebopride, antidopaminergic gastrointestinal prokinetics, on cardiac repolarization.
Effect of clebopride, antidopaminergic gastrointestinal prokinetics, on cardiac repolarization.
International journal of toxicology (2007-03-17)
Ki-Suk Kim, Won-Ho Shin, Sang-joon Park, Eun-Joo Kim
ABSTRACT
The inhibition of the potassium current I(Kr) and QT prolongation has been known to be associated with drug-induced torsades de pointes arrhythmias (TdP) and sudden cardiac death. In this study, the authors investigated the cardiac electrophysiological effects of clebopride, a class of antidopaminergic gastrointestinal prokinetic, that has been reported to prolong the QT interval by using the conventional microelectrode recording techniques in isolated rabbit Purkinje fiber and whole-cell patch clamp techniques in human ether-à-go-go-related gene (hERG)-stably transfected Chinese hamster ovarian (CHO) cells. Clebopride at 10 microM significantly decreased the Vmax of phase 0 depolarization (p < .05) and significantly prolonged the action potential duration at 90% repolarization (APD90) (p < .01), whereas the action potential duration at 50% repolarization (APD50) was not prolonged. For hERG potassium channel currents, the IC50 value was 0.62 +/- 0.30 microM. Clebopride was found to have no effect on sodium channel currents. When these results were compared with Cmax (1.02 nM) of clinical dosage (1 mg, [p.o.]), it can be suggested that clebopride is safe at the clinical dosage of 1 mg from the electrophysiological aspect. These findings indicate that clebopride, an antidopaminergic gastrointestinal prokinetic drug, may provide a sufficient "safety factor" in terms of the electrophysiological threshold concentration. But, in a supratherapeutic concentration that might possibly be encountered during overdose or impaired metabolism, clebopride may have torsadogenic potency.