Merck
Novel anticancer prodrugs of butyric acid. 2.
Journal of medicinal chemistry (1992-02-21)
A Nudelman, M Ruse, A Aviram, E Rabizadeh, M Shaklai, Y Zimrah, A Rephaeli
ABSTRACT
The antitumor activity of novel prodrugs butyric acid was examined. The in vitro effect of the compounds on induction of cytodifferentiation and on inhibition of proliferation and clonogenicity showed that (pivaloyloxy)methyl butyrate (1a) (labeled AN-9) was the most active agent. SAR's suggested that its activity stemmed from hydrolytically released butyric acid. In vivo, 1a displayed antitumor activity in B16F0 melanoma primary cancer model, manifested by a significant increase in the life span of the treated animals. Murine lung tumor burden, induced by injection of the highly metastatic melanoma cells (B16F10.9), was decreased by 1a. It also displayed a significant therapeutic activity against spontaneous metastases which were induced by 3LL Lewis lung carcinoma cells. Moreover, 1a has the advantage of low toxicity, with an acute LD50 = 1.36 +/- 0.1 g/kg (n = 5). These results suggest that 1a is a potential antineoplastic agent.