- Effects of benzylpiperazine derivatives on the neurotoxicity of 3,4-methylenedioxymethamphetamine in rat brain.
Effects of benzylpiperazine derivatives on the neurotoxicity of 3,4-methylenedioxymethamphetamine in rat brain.
Brain research (1992-09-11)
K Hashimoto, H Maeda, T Goromaru
PMID1384935
ABSTRACT
The neurotoxicity of 3,4-methylenedioxymethamphetamine (MDMA) in rat brain was attenuated significantly by coadministration of several benzylpiperazines (p-nitrobenzylpiperazine, p-chlorobenzylpiperazine and 1-piperonylpiperazine), which were weak inhibitors for [3H]6-nitroquipazine binding to the 5-hydroxytryptamine (5-HT) transporter in rat brain. These results suggest that these benzylpiperazines may inhibit the MDMA-induced neurotoxicity by a novel neuropharmacological effect other than 5-HT uptake inhibition.