Merck
  • Enzymatic preparation of genuine prosapogenin, 20(S)-ginsenoside Rh1, from ginsenosides Re and Rg1.
Enzymatic preparation of genuine prosapogenin, 20(S)-ginsenoside Rh1, from ginsenosides Re and Rg1.
Bioscience, biotechnology, and biochemistry (2001-02-24)
S R Ko, Y Suzuki, K J Choi, Y H Kim
ABSTRACT
It was found that a lactase preparation from Penicillium sp. nearly quantitatively hydrolyzed ginsenosides Re and Rg1, which are major saponins in roots of Panax ginseng, to a minor saponin, 20(S)-ginsenoside Rh1 [6-O-beta-D-glucopyranosyl-20(S)-protopanaxatriol]. This is the first report on the enzymatic preparation of ginsenoside Rh1 with a high efficiency. This enzyme also readily hydrolyzed ginsenoside Rg2 to ginsenoside Rh1.
Product Description
Supelco
Ginsenoside Rg2, analytical standard