Merck
  • Treatment of oilfield wastewater in moving bed biofilm reactors using a novel suspended ceramic biocarrier.
Treatment of oilfield wastewater in moving bed biofilm reactors using a novel suspended ceramic biocarrier.
Journal of hazardous materials (2011-09-20)
Zhiyong Dong, Mang Lu, Wenhui Huang, Xiaochun Xu
ABSTRACT
In this study, a novel suspended ceramic carrier was prepared, which has high strength, optimum density (close to water), and high porosity. Two different carriers, unmodified and sepiolite-modified suspended ceramic carriers were used to feed two moving bed biofilm reactors (MBBRs) with a filling fraction of 50% to treat oilfield produced water. The hydraulic retention time (HRT) was varied from 36 to 10h. The results, during a monitoring period of 190 days, showed that removal efficiency of chemical oxygen demand was the highest in reactor 3 filled with the sepiolite-modified carriers, followed by reactor 2 filled with the unmodified carriers, with the lowest in reactor 1 (activated sludge reactor), at an HRT of 10h. Similar trends were found in the removal efficiencies of ammonia nitrogen and polycyclic aromatic hydrocarbons. Reactor 3 was more shock resistant than reactors 2 and 1. The results indicate that the suspended ceramic carrier is an excellent MBBR carrier.
Sigma-Aldrich
Sepiolite, powder