Merck
  • Gel electrophoretic detection of 7,8-dihydro-8-oxoguanine and 7, 8-dihydro-8-oxoadenine via oxidation by Ir (IV).
Gel electrophoretic detection of 7,8-dihydro-8-oxoguanine and 7, 8-dihydro-8-oxoadenine via oxidation by Ir (IV).
Nucleic acids research (1998-06-20)
J G Muller, V Duarte, R P Hickerson, C J Burrows
ABSTRACT
Two gel electrophoretic methods are described for detection of 7, 8-dihydro-8-oxoguanine and 7,8-dihydro-8-oxoadenine based on their further oxidation with one-electron oxidants including IrCl62-and IrBr62-. The products of nucleobase oxidation lead to enhanced piperidine-sensitive cleavage and to highly visible stop points in a primer extension assay. 8-oxoG and 8-oxoA lesions may be distinguished by the latter's inability to be oxidized by IrBr62-compared to IrCl62-Comparison is also made to oxidation by MnO4-.
Sigma-Aldrich
Potassium hexachloroiridate(IV), technical grade
Sigma-Aldrich
Potassium hexachloroiridate(IV), 99.99% trace metals basis