Merck
  • Further study on the use of uncharged beta-cyclodextrin polymer in capillary electrophoresis: enantiomeric separation of some alpha-hydroxy acids.
Further study on the use of uncharged beta-cyclodextrin polymer in capillary electrophoresis: enantiomeric separation of some alpha-hydroxy acids.
Electrophoresis (1995-08-01)
S Fanali, Z Aturki
ABSTRACT
Uncharged beta-cyclodextrin polymer was used as chiral selector for the enantiomeric separation of some alpha-hydroxy acids by capillary electrophoresis. Complexation and enantiomeric resolution of mandelic acid, m-hydroxy and p-hydroxymandelic acid, 3,4-dihydroxymandelic acid, as well as 2- and 3-phenyllactic acid were studied, changing the concentration of the beta-cyclodextrin polymer added to the background electrolyte at different pH in the range of 4.5-7. Furthermore, the effects of the concentration of the background electrolyte, column temperature, and applied voltage on chiral resolution were also examined. The best enantiomeric separations were obtained using a background electrolyte at pH 6 containing 100 mg/mL of beta-cyclodextrin polymer.
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%
Sigma-Aldrich
3-Hydroxymandelic acid, ≥97.0% (T)