- 5-HT1A and D-2 receptor affinity of o-methoxyphenylpiperazine derivatives with terminal benzamide fragment on N-4 alkyl chain. 2.
5-HT1A and D-2 receptor affinity of o-methoxyphenylpiperazine derivatives with terminal benzamide fragment on N-4 alkyl chain. 2.
Farmaco (Societa chimica italiana : 1989) (1995-07-01)
R Perrone, F Berardi, M Leopoldo, V Tortorella, M D Lograno, E Daniele, S Govoni
PMID7669188
ABSTRACT
The synthesis of some o-methoxyphenylpiperazines with a benzamide moiety on N-4 alkyl chain was accomplished and their affinity for dopamine and serotonin receptor subtypes was assayed by in vitro receptor binding. The results show that several derivatives had a good affinity both on D-2 and 5-HT1A receptors, the IC50s ranging from 10(-7) to 10(-8) M.
MATERIALS