Merck
  • Active site structural change of alpha-chymotrypsin due to 2-halogeno-ethanols; comparison with ethanol, I-propanol, and urea.
Active site structural change of alpha-chymotrypsin due to 2-halogeno-ethanols; comparison with ethanol, I-propanol, and urea.
Experientia (1980-05-15)
S Ebina, Y Nagai
ABSTRACT
2-Halogeno-ethanols change the active site structure of alpha-chymotrypsin more rapidly and effectively than ethanol, 1-propanol and urea, probably before producing an extensive conformation change.
Sigma-Aldrich
2-Bromoethanol, 95%
Supelco
2-Bromoethanol, analytical standard