Merck
  • Kinetic study on the slow inhibition of epidermis tyrosinase by m-coumaric acid.
Kinetic study on the slow inhibition of epidermis tyrosinase by m-coumaric acid.
Biochimica et biophysica acta (1984-10-23)
J Cabanes, F García-Carmona, F García-Cánovas, J L Iborra, J A Lozano
ABSTRACT
The inhibition by m-coumaric acid of oxidation of L-dopa by epidermis tyrosinase (monophenol,dihydroxy-L-phenylalanine:oxygen oxidoreductase, EC 1.14.18.1) is characterized by a prolonged transient phase. Kinetic data correspond to that for a postulated mechanism that involves rapid formation of a reduced enzyme-m-coumaric acid complex that subsequently undergoes a relatively slow reversible reaction. An overall inhibition constant for m-coumaric acid of 0.05 mM was calculated. The value of the Ki for the dissociation of m-coumaric acid from the rapidly formed complex was calculated as 0.53 mM. The first-order rate constants for the slow isomerization of the enzyme-inhibitor complex were calculated as 3.0 +/- 0.1 min-1 for the forward step and 0.31 +/- 0.06 min-1 for the reverse step.
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Supelco
m-Coumaric acid, analytical standard