Merck
  • Metabolism of volatile chlorinated aliphatic hydrocarbons by Pseudomonas fluorescens.
Metabolism of volatile chlorinated aliphatic hydrocarbons by Pseudomonas fluorescens.
Applied and environmental microbiology (1988-10-01)
P A Vandenbergh, B S Kunka
ABSTRACT
A Pseudomonas fluorescens strain designated PFL12 was isolated from soil and water that were contaminated with various chloroaliphatic hydrocarbons. The isolate was able to metabolize 1,2-dichloroethane, 1,1,2-trichloroethane, 1,2-dichloropropane, 2,2-dichloropropane, and trichloroethylene.
Product Description
Supelco
Headspace vial, screw top, rounded bottom (vial only), volume 20 mL, clear glass vial, thread for 18, O.D. × H 22.5 mm × 75.5 mm, pkg of 100 ea
Supelco
Headspace vial, screw top, rounded bottom (vial only), volume 10 mL, clear glass vial, thread for 18, O.D. × H 22.5 mm × 46 mm, pkg of 100 ea
Sigma-Aldrich
1,1,2-Trichloroethane, contains ≤3% 2-propanol as stabilizer, 97%
Supelco
1,1,2-Trichloroethane, analytical standard