Merck
  • Cognitive enhancement effects of Bacopa monnieri (Brahmi) on novel object recognition and NMDA receptor immunodensity in the prefrontal cortex and hippocampus of sub-chronic phencyclidine rat model of schizophrenia.
Cognitive enhancement effects of Bacopa monnieri (Brahmi) on novel object recognition and NMDA receptor immunodensity in the prefrontal cortex and hippocampus of sub-chronic phencyclidine rat model of schizophrenia.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-08-14)
Pritsana Piyabhan, Thanitsara Wetchateng, Seewaboon Sireeratawong
ABSTRACT
Cognitive impairment is a common characteristic in schizophrenia that cannot be attenuated by antipsychotics. Brahmi, popularly known as a cognitive enhancer might be a new frontier of cognitive deficit treatment in schizophrenia. To study effects of Brahmi on attenuation at cognitive deficit and cerebral glutamate/N-methyl-D-aspartate (NMDA) receptor density in sub-chronic phencyclidine (PCP) rat model of schizophrenia. Rats were administered PCP or vehicle. Half of the PCP-group was treated with Brahmi. Discrimination ratio (DR) representing cognitive ability was obtained from novel object recognition task. NMDA immunodensity was measured in prefrontal cortex, striatum, cornu ammonis fields 1 to 3 of hippocampus (CA1-3), and dentate gyrus (DG) using immunohistochemistry. DR in PCP-group was significantly decreased compared with control. This occurred alongside NMDA up-regulation in prefrontal cortex and CA1-3, but not in striatum and DG. PCP with Brahmi showed a significant increase in DR score compared with PCP alone. This occurred alongside significant decrease in NMDA immunodensity in prefrontal cortex and CA1-3. No significant difference in cerebral NMDA immunodensity was observed between PCP with Brahmi and control. Cognitive deficit observed in PCP-administered rats was mediated by NMDA up-regulation in prefrontal cortex and CA1-3. Interestingly, Brahmi could recover this cognitive deficit by decreasing NMDA density in these brain areas to normal.
Product Description
Supelco
PCP (Phencyclidine) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®