Merck
  • Mineralocorticoid receptor agonists induce mouse aortic aneurysm formation and rupture in the presence of high salt.
Mineralocorticoid receptor agonists induce mouse aortic aneurysm formation and rupture in the presence of high salt.
Arteriosclerosis, thrombosis, and vascular biology (2013-05-11)
Shu Liu, Zhongwen Xie, Alan Daugherty, Lisa A Cassis, Kevin J Pearson, Ming C Gong, Zhenheng Guo
ABSTRACT
Elevated plasma aldosterone concentrations in patients have been linked to a spectrum of cardiovascular diseases. Mineralocorticoid receptor antagonists provide additional benefits in patients with heart failure. However, whether aldosterone and the mineralocorticoid receptor are involved in aortic aneurysm is unknown. We report that administration of deoxycorticosterone acetate (DOCA) and salt or aldosterone and salt, but not DOCA or salt alone, to C57BL/6 male mice induced abdominal and thoracic aortic aneurysm formation and rupture in an age-dependent manner. DOCA and salt- or aldosterone and salt-induced aortic aneurysm mimicked human aortic aneurysm with respect to elastin degradation, inflammatory cell infiltration, smooth muscle cell degeneration and apoptosis, and oxidative stress. Aortic aneurysm formation did not correlate with the increase in blood pressure induced by DOCA and salt. Systemic administration of the angiotensin-converting enzyme inhibitor, enalapril, or angiotensin type 1 receptor antagonist, losartan, did not affect DOCA and salt-induced aortic aneurysm. In contrast, the mineralocorticoid receptor antagonists, spironolactone or eplerenone, significantly attenuated DOCA and salt- or aldosterone and salt-induced aortic aneurysm. The current study describes a novel aortic aneurysm animal model induced by mineralocorticoid receptor agonist and high salt, and reveals a previously unrecognized but potentially significant role of aldosterone in the pathogenesis of aortic aneurysm. These findings imply that mineralocorticoid receptor antagonists may be effective in the treatment of some aortic aneurysms.
Product Description
Spironolactone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Supelco
Losartan potassium
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Losartan potassium, European Pharmacopoeia (EP) Reference Standard
Supelco
Aldosterone solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Supelco
Enalapril Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Enalapril maleate salt, powder, ≥98% (TLC)