Merck
  • Effects of Copaifera duckei Dwyer oleoresin on the cell wall and cell division of Bacillus cereus.
Effects of Copaifera duckei Dwyer oleoresin on the cell wall and cell division of Bacillus cereus.
Journal of medical microbiology (2013-05-04)
Elizabeth Cristina Gomes Dos Santos, Claudio Luis Donnici, Elizabeth Ribeiro da Silva Camargos, Adriana Augusto de Rezende, Eloisa Helena de Aguiar Andrade, Luiz Alberto Lira Soares, Luiz de Macêdo Farias, Maria Auxiliadora Roque de Carvalho, Maria das Graças Almeida
ABSTRACT
The aim of this work was to evaluate the antibacterial activity of Copaifera duckei oleoresin and to determine its possible mechanism of action against bacteria of clinical and food interest. The antibacterial activity was determined by agar diffusion and dilution methods; the mechanism of action by transmission electron microscopy and by SDS-PAGE; the bioactive compounds by bioautography; and the chemical analysis by GC/MS. Oleoresin showed activity against nine of the 11 strains of bacteria tested. Bacillus cereus was the most sensitive, with a MIC corresponding to 0.03125 mg ml(-1) and with a bactericidal action. Oleoresin acted on the bacterial cell wall, removing proteins and the S-layer, and interfering with the cell-division process. This activity probably can be attributed to the action of terpenic compounds, among them the bisabolene compound. Gram-negative bacteria tested were not inhibited. C. duckei oleoresin is a potential antibacterial, suggesting that this oil could be used as a therapeutic alternative, mainly against B. cereus.
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)
Millipore
Vancomycin supplement, suitable for microbiology