Merck
  • A retrospective audit to evaluate the effect of the change in midazolam concentration from 10 mg/5 ml to 5 mg/5 ml ampoules on midazolam doses given to sedation patients following the rapid response report (RRR).
A retrospective audit to evaluate the effect of the change in midazolam concentration from 10 mg/5 ml to 5 mg/5 ml ampoules on midazolam doses given to sedation patients following the rapid response report (RRR).
SAAD digest (2013-04-02)
Shazia Kaka, Pan Taneja
ABSTRACT
The technique of intravenous sedation with a benzodiazepine is a well documented and successful method for reducing fear and anxiety in many patients who regard dental treatment as challenging and potentially painful. One of the most important factors to achieving a well sedated patient is ensuring the dose of the drug given is titrated to the individual patient. Five years ago, a Rapid Response Report issued by the National Patient Safety Agency changed the concentration of midazolam within a 5 ml ampoule from 10 mg/5 ml to 5 mg/5 ml. A retrospective audit of 300 patients undergoing oral surgery treatment under intravenous midazolam sedation at Birmingham Dental Hospital was carried out over a 6-month period to assess whether this difference in midazolam concentration had any influence on the average dose of midazolam given to these patients. The midazolam doses given by five clinicians were audited both before and after the concentration change. the mean midazolam dose that was administered by three clinicians decreased following the change in concentration. In addition, the number of patients given greater than 5 mg midazolam following the change decreased with all five clinicians. The change in midazolam concentration from 10 mg/5 ml to 5 mg/5 ml ampoules has resulted in the overall average dose of midazolam given by five clinicians at Birmingham Dental Hospital to decrease.
Product Description
Supelco
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®