Merck
  • Regulation of IL-17A production is distinct from IL-17F in a primary human cell co-culture model of T cell-mediated B cell activation.
Regulation of IL-17A production is distinct from IL-17F in a primary human cell co-culture model of T cell-mediated B cell activation.
PloS one (2013-03-19)
Andrew C Melton, Jennifer Melrose, Liisa Alajoki, Sylvie Privat, Hannah Cho, Naomi Brown, Ana Marija Plavec, Dat Nguyen, Elijah D Johnston, Jian Yang, Mark A Polokoff, Ivan Plavec, Ellen L Berg, Alison O'Mahony
ABSTRACT
Improper regulation of B cell responses leads to excessive production of antibodies and contributes to the development of autoimmune disease. T helper 17 (Th17) cells also drive the development of autoimmune disease, but the role of B cells in shaping Th17 cell-mediated immune responses, as well as the reciprocal regulation of B cell responses by IL-17 family cytokines, remains unclear. The aim of this study was to characterize the regulation of IL-17A and IL-17F in a model of T cell-dependent B cell activation. Stimulation of primary human B cell and peripheral blood mononuclear cell (BT) co-cultures with α-IgM and a non-mitogenic concentration of superantigens for three days promoted a Th17 cell response as evidenced by increased expression of Th17-related gene transcripts, including Il17f, Il21, Il22, and Il23r, in CD4 T cells, as well as the secretion of IL-17A and IL-17F protein. We tested the ability of 144 pharmacologic modulators representing 91 different targets or pathways to regulate IL-17A and IL-17F production in these stimulated BT co-cultures. IL-17A production was found to be preferentially sensitive to inhibition of the PI3K/mTOR pathway, while prostaglandin EP receptor agonists, including PGE2, increased IL-17A concentrations. In contrast, the production of IL-17F was inhibited by PGE2, but selectively increased by TLR2 and TLR5 agonists. These results indicate that IL-17A regulation is distinct from IL-17F in stimulated BT co-cultures and that this co-culture approach can be used to identify pathway mechanisms and novel agents that selectively inhibit production of IL-17A or IL-17F.
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Supelco
1α,25-Dihydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®