Merck
  • Integrated system for temperature-controlled fast protein liquid chromatography comprising improved copolymer modified beaded agarose adsorbents and a travelling cooling zone reactor arrangement.
Integrated system for temperature-controlled fast protein liquid chromatography comprising improved copolymer modified beaded agarose adsorbents and a travelling cooling zone reactor arrangement.
Journal of chromatography. A (2013-03-14)
Tobias K H Müller, Ping Cao, Stephanie Ewert, Jonas Wohlgemuth, Haiyang Liu, Thomas C Willett, Eirini Theodosiou, Owen R T Thomas, Matthias Franzreb
ABSTRACT
An integrated approach to temperature-controlled chromatography, involving copolymer modified agarose adsorbents and a novel travelling cooling zone reactor (TCZR) arrangement, is described. Sepharose CL6B was transformed into a thermoresponsive cation exchange adsorbent (thermoCEX) in four synthetic steps: (i) epichlorohydrin activation; (ii) amine capping; (iii) 4,4'-azobis(4-cyanovaleric acid) immobilization; and 'graft from' polymerization of poly(N-isopropylacrylamide-co-N-tert-butylacrylamide-co-acrylic acid-co-N,N'-methylenebisacrylamide). FT-IR, (1)H NMR, gravimetry and chemical assays allowed precise determination of the adsorbent's copolymer composition and loading, and identified the initial epoxy activation step as a critical determinant of 'on-support' copolymer loading, and in turn, protein binding performance. In batch binding studies with lactoferrin, thermoCEX's binding affinity and maximum adsorption capacity rose smoothly with temperature increase from 20 to 50 °C. In temperature shifting chromatography experiments employing thermoCEX in thermally jacketed columns, 44-51% of the lactoferrin adsorbed at 42 °C could be desorbed under binding conditions by cooling the column to 22 °C, but the elution peaks exhibited strong tailing. To more fully exploit the potential of thermoresponsive chromatography adsorbents, a new column arrangement, the TCZR, was developed. In TCZR chromatography, a narrow discrete cooling zone (special assembly of copper blocks and Peltier elements) is moved along a bespoke fixed-bed separation columnfilled with stationary phase. In tests with thermoCEX, it was possible to recover 65% of the lactoferrin bound at 35 °C using 8 successive movements of the cooling zone at a velocity of 0.1mm/s; over half of the recovered protein was eluted in the first peak in more concentrated form than in the feed. Intra-particle diffusion of desorbed protein out of the support pores, and the ratio between the velocities of the cooling zone and mobile phase were identified as the main parameters affecting TCZR performance. In contrast to conventional systems, which rely on cooling the whole column to effect elution and permit only batch-wise operation, TCZR chromatography generates sharp concentrated elution peaks without tailing effects and appears ideally suited for continuous operation.
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Sepharose CL-6B, Cross-linked